Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cycp3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130072.3 | NP_182034.1 | 1 - 2 | 449 | 130 | |
GenBank mRNA | AY074650.1 | - | 1 - 2 | 412 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |