Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337118.1 | NP_001325142.1 | 12 - 13 | 1063 | 69 | |
NM_130087.4 | NP_182049.1 | 12 - 13 | 1189 | 69 | ||
GenBank mRNA | AF250960.1 | - | 12 - 13 | 945 | 69 | |
AK227222.1 | - | 12 - 13 | 985 | 69 | ||
AY099542.1 | - | 12 - 13 | 960 | 69 | ||
BT008490.1 | - | 12 - 13 | 890 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |