Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202820.1 | NP_001189749.1 | 2 - 3 | 429 | 66 | |
NM_130091.2 | NP_566040.1 | 2 - 3 | 357 | 66 | ||
GenBank mRNA | AB062456.1 | - | 2 - 3 | 357 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |