Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202824.1 | NP_001189753.1 | 3 - 4 | 404 | 64 | |
NM_130126.4 | NP_566050.1 | 3 - 4 | 316 | 64 | ||
GenBank mRNA | AF378876.1 | - | 3 - 4 | 271 | 64 | |
AK227453.1 | - | 3 - 4 | 267 | 64 | ||
AY050474.1 | - | 3 - 4 | 256 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |