Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130132.4 | NP_566053.1 | 1 - 2 | 228 | 125 | |
GenBank mRNA | AK175396.1 | - | 1 - 2 | 193 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |