Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130132.4 | NP_566053.1 | 3 - 4 | 502 | 91 | |
GenBank mRNA | AK175396.1 | - | 3 - 4 | 467 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |