Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130142.4 | NP_182103.1 | 1 - 2 | 313 | 69 | |
GenBank mRNA | AY050806.1 | - | 1 - 2 | 200 | 69 | |
AY088930.1 | - | 1 - 2 | 209 | 69 | ||
AY113964.1 | - | 1 - 2 | 112 | 69 | ||
DQ442991.1 | - | 1 - 2 | 112 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |