Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337186.1 | NP_001325255.1 | 6 - 7 | 3442 | 74 | |
NM_130197.5 | NP_182157.2 | 6 - 7 | 3263 | 74 | ||
GenBank mRNA | AF135455.1 | - | 6 - 7 | 3036 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |