Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036472.2 | NP_001031549.1 | 2 | 1337 | 149 | |
NM_130224.8 | NP_182182.2 | 1 | 2047 | 149 | ||
GenBank mRNA | BT003328.1 | - | 1 | 1743 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |