Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G47320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130300.4 | NP_182254.1 | 2 - 3 | 270 | 98 | |
GenBank mRNA | AY568520.1 | - | 1 - 2 | 157 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |