Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G47320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130300.4 | NP_182254.1 | 3 - 4 | 310 | 72 | |
GenBank mRNA | AY568520.1 | - | 2 - 3 | 197 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |