Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130311.2 | NP_182265.1 | 5 - 6 | 489 | 87 | |
GenBank mRNA | D87545.1 | - | 5 - 6 | 489 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |