Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130326.3 | NP_182280.1 | 2 - 3 | 311 | 101 | |
GenBank mRNA | AY059875.1 | - | 2 - 3 | 311 | 101 | |
AY093341.1 | - | 2 - 3 | 269 | 101 | ||
Z49991.1 | - | 298 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |