Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130328.3 | NP_566105.1 | 7 - 8 | 1524 | 91 | |
GenBank mRNA | AF109178.1 | - | 7 - 8 | 1357 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |