Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130347.5 | NP_182301.1 | 2 - 3 | 2697 | 75 | |
GenBank mRNA | AF243509.1 | - | 2 - 3 | 2419 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |