Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130365.4 | NP_566119.1 | 14 - 15 | 2306 | 73 | |
GenBank mRNA | AY063915.1 | - | 14 - 15 | 2205 | 73 | |
AY091270.1 | - | 14 - 15 | 2090 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |