Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331724.1 | NP_001322003.1 | 1 | 1643 | 137 | |
NM_100646.3 | NP_172251.1 | 1 | 1515 | 137 | ||
GenBank mRNA | AF428349.1 | - | 1 | 1468 | 137 | |
AY062700.1 | - | 1 | 1468 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |