Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331741.1 | NP_001318950.1 | 18 - 19 | 2558 | 66 | |
NM_100665.3 | NP_172269.2 | 18 - 19 | 2436 | 66 | ||
GenBank mRNA | AK230326.1 | - | 18 - 19 | 2403 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |