Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331741.1 | NP_001318950.1 | 26 - 27 | 3287 | 117 | |
NM_100665.3 | NP_172269.2 | 26 - 27 | 3165 | 117 | ||
GenBank mRNA | AK230326.1 | - | 26 - 27 | 3132 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |