Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100681.2 | NP_172285.2 | 5 - 6 | 806 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |