Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100689.5 | NP_172293.2 | 16 - 17 | 2431 | 65 | |
GenBank mRNA | AK117238.1 | - | 16 - 17 | 2439 | 65 | |
BT005964.1 | - | 16 - 17 | 2300 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |