Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331765.1 | NP_001322767.1 | 1 - 2 | 2246 | 64 | |
NM_100699.6 | NP_172303.5 | 1 - 2 | 2246 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |