Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331765.1 | NP_001322767.1 | 5 - 6 | 2630 | 91 | |
NM_100699.6 | NP_172303.5 | 5 - 6 | 2630 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |