Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 11 - 12 | 1935 | 69 | |
GenBank mRNA | AK226535.1 | - | 11 - 12 | 1820 | 69 | |
AX765812.1 | - | 11 - 12 | 1778 | 69 | ||
AY063821.1 | - | 11 - 12 | 1836 | 69 | ||
AY091402.1 | - | 11 - 12 | 1778 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |