Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 1 - 2 | 429 | 57 | |
GenBank mRNA | AK226535.1 | - | 1 - 2 | 314 | 57 | |
AX765812.1 | - | 1 - 2 | 272 | 57 | ||
AY063821.1 | - | 1 - 2 | 330 | 57 | ||
AY091402.1 | - | 1 - 2 | 272 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |