Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 5 - 6 | 1132 | 141 | |
GenBank mRNA | AK226535.1 | - | 5 - 6 | 1017 | 141 | |
AX765812.1 | - | 5 - 6 | 975 | 141 | ||
AY063821.1 | - | 5 - 6 | 1033 | 141 | ||
AY091402.1 | - | 5 - 6 | 975 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |