Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 9 - 10 | 1676 | 64 | |
GenBank mRNA | AK226535.1 | - | 9 - 10 | 1561 | 64 | |
AX765812.1 | - | 9 - 10 | 1519 | 64 | ||
AY063821.1 | - | 9 - 10 | 1577 | 64 | ||
AY091402.1 | - | 9 - 10 | 1519 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |