Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331787.1 | NP_001321098.1 | 24 - 25 | 3477 | 135 | |
NM_100746.3 | NP_172349.2 | 24 - 25 | 2976 | 135 | ||
GenBank mRNA | LC177512.1 | - | 23 - 24 | 2952 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |