Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331787.1 | NP_001321098.1 | 31 - 32 | 4548 | 145 | |
NM_100746.3 | NP_172349.2 | 31 - 32 | 4047 | 145 | ||
GenBank mRNA | LC177512.1 | - | 30 - 31 | 4023 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |