Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331787.1 | NP_001321098.1 | 37 - 38 | 4993 | 112 | |
NM_100746.3 | NP_172349.2 | 37 - 38 | 4492 | 112 | ||
GenBank mRNA | LC177512.1 | - | 36 - 37 | 4468 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |