Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAP27-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035927.2 | NP_001031004.1 | 6 - 7 | 875 | 67 | |
NM_001331791.1 | NP_001321657.1 | 6 - 7 | 1468 | 67 | ||
NM_001331792.1 | NP_001321655.1 | 6 - 7 | 1314 | 67 | ||
NM_001331793.1 | NP_001321656.1 | 6 - 7 | 1250 | 67 | ||
NM_100756.2 | NP_172359.2 | 7 - 8 | 793 | 67 | ||
GenBank mRNA | AK317489.1 | - | 6 - 7 | 648 | 67 | |
AK317626.1 | - | 7 - 8 | 794 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |