Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035929.1 | NP_001031006.1 | 14 - 15 | 1279 | 78 | |
NM_100765.5 | NP_563830.1 | 14 - 15 | 1444 | 78 | ||
GenBank mRNA | AJ249967.1 | - | 14 - 15 | 1236 | 78 | |
BT008661.1 | - | 14 - 15 | 1254 | 78 | ||
D89051.1 | - | 14 - 15 | 1179 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |