Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100779.5 | NP_172382.4 | 1 - 2 | 113 | 88 | |
GenBank mRNA | AY156728.1 | - | 1 - 2 | 113 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |