Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198015.2 | NP_001184944.1 | 2 - 3 | 391 | 94 | |
NM_100779.5 | NP_172382.4 | 2 - 3 | 412 | 94 | ||
GenBank mRNA | AY156728.1 | - | 2 - 3 | 412 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |