Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100825.5 | NP_172425.2 | 2 - 3 | 497 | 151 | |
GenBank mRNA | AY063939.1 | - | 2 - 3 | 437 | 151 | |
AY096523.1 | - | 2 - 3 | 265 | 151 | ||
AY519552.1 | - | 2 - 3 | 265 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |