Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100859.3 | NP_563855.1 | 3 - 4 | 963 | 77 | |
GenBank mRNA | AK226753.1 | - | 3 - 4 | 825 | 77 | |
BT026490.1 | - | 3 - 4 | 800 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |