Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100867.2 | NP_172464.1 | 4 - 5 | 1607 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |