Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035938.2 | NP_001031015.1 | 2 - 3 | 149 | 76 | |
NM_100881.3 | NP_172478.1 | 1 - 2 | 233 | 76 | ||
GenBank mRNA | AF370135.1 | - | 1 - 2 | 230 | 76 | |
AJ271731.1 | - | 1 - 2 | 117 | 76 | ||
AK317528.1 | - | 2 - 3 | 150 | 76 | ||
AY051038.1 | - | 1 - 2 | 65 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |