Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100894.4 | NP_172491.1 | 1 - 2 | 517 | 74 | |
GenBank mRNA | AF370569.1 | - | 1 - 2 | 141 | 74 | |
AY085377.1 | - | 1 - 2 | 225 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |