Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100919.5 | NP_172515.2 | 19 - 20 | 3737 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |