Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331912.1 | NP_001320848.1 | 11 - 12 | 1515 | 119 | |
NM_100929.5 | NP_172524.1 | 12 - 13 | 1517 | 119 | ||
GenBank mRNA | AY063935.1 | - | 12 - 13 | 1264 | 119 | |
AY096464.1 | - | 12 - 13 | 1245 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |