Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100930.4 | NP_172525.1 | 2 - 3 | 402 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |