Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331933.1 | NP_001320312.1 | 18 - 19 | 2418 | 71 | |
NM_100962.4 | NP_172556.2 | 18 - 19 | 2403 | 71 | ||
GenBank mRNA | AJ130878.1 | - | 18 - 19 | 2338 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |