Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQ4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100968.3 | NP_172562.2 | 9 - 10 | 1768 | 119 | |
GenBank mRNA | AY120761.1 | - | 9 - 10 | 1666 | 119 | |
BT010133.1 | - | 8 - 9 | 1462 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |