Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100991.2 | NP_172585.1 | 4 - 5 | 551 | 83 | |
GenBank mRNA | AY040016.1 | - | 4 - 5 | 551 | 83 | |
AY079368.1 | - | 4 - 5 | 467 | 83 | ||
AY088393.1 | - | 4 - 5 | 551 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |