Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125011.4 | NP_568839.1 | 1 - 2 | 371 | 153 | |
GenBank mRNA | BT025877.1 | - | 1 - 2 | 296 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |