Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G56730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345214.1 | NP_001318814.1 | 5 - 6 | 890 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |