Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345216.1 | NP_001330516.1 | 4 - 5 | 595 | 70 | |
NM_125058.3 | NP_200486.1 | 4 - 5 | 711 | 70 | ||
GenBank mRNA | AY050428.1 | - | 4 - 5 | 463 | 70 | |
AY087544.1 | - | 4 - 5 | 463 | 70 | ||
AY090332.1 | - | 4 - 5 | 354 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |