Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125079.3 | NP_200507.1 | 4 - 5 | 1320 | 69 | |
GenBank mRNA | AF303979.1 | - | 4 - 5 | 1273 | 69 | |
BT033021.1 | - | 4 - 5 | 1273 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |