Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125104.3 | NP_200532.1 | 2 - 3 | 936 | 70 | |
GenBank mRNA | AY065209.1 | - | 2 - 3 | 937 | 70 | |
AY096511.1 | - | 2 - 3 | 865 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |